Natriumklorid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Leverandør: VWR Chemicals

Synonymer: Saline, NaCl, Sea salt, Halite

Dose for preparation of 1 litre of solution.

32064.600EA 555 NOK
32064.600
Natriumklorid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Natriumklorid
Formel: NaCl
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3

Bestill nå

Spesifikasjon Testresultater

Titer (20°C) 0.0995 - 0.1005 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR