Natriumhydroksid concentrate for dilution to 1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Leverandør: VWR Chemicals
Danger

Synonymer: Caustic soda

Dose for preparation of 1 litre of solution.

32066.606EA 345 NOK
32066.606
Natriumhydroksid concentrate for dilution to 1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Natriumhydroksid
Formel: NaOH
Kokepunkt: >105 °C (1013 hPa)
Tetthet: 1,327 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Titer (20°C) 0.995 to 1.005 mol/l
Conforms to BDH 18042 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR