EDTA solution, 0,5 M, molecular biology grade, Calbiochem®, Millipore®
Leverandør: Merck Millipore Calbiochem
324506-100
324506-100EA 1125 NOK
324506-100
EDTA solution, 0,5 M, molecular biology grade, Calbiochem®, Millipore®
Buffere Buffere, kloning
Sterile-filtered solution of 0,5 M EDTA in H₂O treated with diethyl pyrocarbonate (DEPC). Used in molecular biology applications.
  • pH: 8,0
  • Diethyl pyrocarbonate (DEPC) treated
Synonyms: Ethylenediaminetetraacetic acid trisodium salt.