Pesticide reference standards, Pyridaphenthion, PESTANAL®, analytical standards, Supelco®
Leverandør: Merck
32538-100MG
32538-100MGEA 1275 NOK
32538-100MG
Pesticide reference standards, Pyridaphenthion, PESTANAL®, analytical standards, Supelco®
Standarder Organiske standarder
Pyridaphenthion may be used as a reference standard for the quantification of pyridaphenthion in biological samples using liquid chromatography-atmospheric pressure chemical ionisation mass spectrometry.
  • This insecticide is effective against sucking and chewing insects and spider mites
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef70ebec16e881d9547cb188f98d62ab5811e242&vid=ef70ebec16e881d9547cb188f98d62ab5811e242&prid=web&clid=SAPDAM