2-Methoxy-4-nitrophenol 97%, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

326828-5G
326828-5GEA 1410 NOK
326828-5G
2-Methoxy-4-nitrophenol 97%, Sigma-Aldrich®
2-Methoxy-4-nitrophenol
Formel: C₇H₇NO₄
MW: 169.14 g/mol
Smeltepunkt: 103…104 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012143
CAS-nummer: 3251-56-7
EINECS: 221-839-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=450d7f02f29e1f5dc937ba4fbabf8778956a1c94&vid=1e20f2eb8a1e84e6d2de4401990b534284640469&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Yellow
Appearance (Form) Crystal to Powder
Infrared spectrum Conforms to Structure
Carbon 48.0 - 51.4%
Nitrogen 8.0 - 8.6%
Purity (GC) ≥96.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR