Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Leverandør: Merck
32758-1ML
32758-1MLEA 27550 NOK
32758-1ML
Organic reference standard, Zearalenone-¹³C₁₈ solution, analytical standard, Supelco®
Standarder Organiske standarder
Zearalenone-¹³C₁₈ may be used as an analytical standard for the determination of zearalenone in animal feed by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
Zearalenone-¹³C₁₈ is the ¹³C isotope-labeled analog of the mycotoxin, zearalenone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8843340394c7ccdba9861a50b7580a57bc6d28d&vid=d8843340394c7ccdba9861a50b7580a57bc6d28d&prid=web&clid=SAPDAM