Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Leverandør: Merck
32834-10MG
32834-10MGEA 5400 NOK
32834-10MG
Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Standarder Organiske standarder
Leucocrystal violet-d₆ might be used as an internal standard in the determination of triphenylmethane dyes in rainbow trout samples using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc197898fcfc3e1cc302f78fc363ca825d4f6c5&vid=dcc197898fcfc3e1cc302f78fc363ca825d4f6c5&prid=web&clid=SAPDAM