Giemsa's stain modified solution, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

M-Clarity™ quality level = MQ200

32884-1L 32884-250ML
32884-1LEA 1170 NOK
32884-1L 32884-250ML
Giemsa's stain modified solution, Sigma-Aldrich®
Giemsa's stain

Modified Giemsa stain may be used for the identification of each human chromosome by differentials staining.


Modified Giemsa stain means when the pH of the Giemsa stain is changed from the usual 6.8 to 9.0.

Tetthet: 0.99 g/cm³ (20 °C)
Flash punkt: 11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
EINECS: 257-438-2
FN: 1230
ADR: 3 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR