Organic reference standard, 3-Acetyldeoxynivalenol, Supelco®

Leverandør: Merck

32927-5MG
32927-5MGEA 7025 NOK
32927-5MG
Organic reference standard, 3-Acetyldeoxynivalenol, Supelco®
Standarder Organiske standarder

3-Acetyldeoxynivalenol belongs to the class of trichothecenes, which are secondary metabolites generated by various fungi, particularly by Fusarium species.


3-Acetyldeoxynivalenol may be used as an analytical reference standard for the determination of the analyte in: Cereals by gas chromatography-mass spectrometry (GC-MS), milk samples by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=479183d2e824835572ced0ace2e622d04eaed033&vid=dd5edb6c9a3dbdc0df7f2fc78dea495848acd67d&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse 3-Acetyldeoxynivalenol
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 50722-38-8
Formel C₁₇H₂₂O₇
kokepunkt 130 °C mmHg (lit.)
Flammepunkt 44 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR