Organic reference standard, 15-Acetyldeoxynivalenol, Supelco®
Leverandør: Merck
32928-5MG
32928-5MGEA 6425 NOK
32928-5MG
Organic reference standard, 15-Acetyldeoxynivalenol, Supelco®
Standarder Organiske standarder
15-Acetyldeoxynivalenols belong to the class of type B trichothecene mycotoxins, generated by several Fusarium fungi.
15-Acetyldeoxynivalenol can be used as an analytical reference standard for the determination of the analyte in: Pig urine and maize samples by high performance liquid chromatography-mass spectrometry (HPLC-MS), milk samples by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21e9551036224cbc4c21ae3e7fb77eb0b879ec8a&vid=21e9551036224cbc4c21ae3e7fb77eb0b879ec8a&prid=web&clid=SAPDAM