Resazurin sodium salt, tablets for milk analysis

Leverandør: VWR Chemicals

Synonymer: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

330884YEA 685 NOK
330884Y
Resazurin sodium salt, tablets for milk analysis
Resazurin sodium salt

A standard solution of resazurin is prepared by dissolving one tablet in 50ml of sterile glass-distilled water. One ml of this solution should be added to 10ml of milk for each test. Solutions must be made freshly each day.


During incubation with milk resazurin undergoes reduction and the colour changes from blue to pink to colourless. The rate of colour change is an indication of the number of microbial organisms in the sample.

Formel: C₁₂H₆NNaO₄
MW: 251.17 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8
EINECS: 263-718-5

Bestill nå

Spesifikasjon Testresultater

Appearance Red to purple tablets
Shape Round biconvex
Average tablet weight 240 - 260 mg
Diameter 7.50 - 8.00 mm
Solubility in water (2 tablets/100 ml) Max. 4.0 min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR