Skje
Leverandør: BURKLE
5354-1000
331-0056EA 9425 NOK
331-0056
Skje
Øser
Stainless steel V2A (1.4301).
  • Complete with rod
  • No openings, undercuts or crevices in which residues could collect
  • Pore-free finish produced by high quality laser welding seams
  • Sterilisable by flame or steam sterilisation