VWR®, flasker, smal hals, Winchester-type, med skrukork
Leverandør: LAB3 OWN PRODUCTION
331-0183EA 21150 NOK
331-0183
VWR®, flasker, smal hals, Winchester-type, med skrukork
Flasker Glassflasker
Glass, clear or amber, with white screw cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Amber coloured to protect light-sensitive materials
Excellent for sampling, storing and transporting liquids.