VWR®, Bottles, Wide Neck, with Screw Cap
Leverandør: VWR Collection
331-0205EA 2600 NOK
331-0205
VWR®, Bottles, Wide Neck, with Screw Cap
Flasker Plastflasker
PE, translucent with black PP caps.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The extra wide mouth allows easy filling and cleaning.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904.