Detectable spoons, long handle, blue, SteriPlast®
Leverandør: BURKLE
5378-5004 5378-5008
331-0331EA 845 NOK
331-0331 331-0332
Detectable spoons, long handle, blue, SteriPlast®
Skjeer
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
  • PS detectable
  • Individually packaged and sterilised, for single use
  • Can be detected by colour scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS/BRC foreign object management