Ladle, long handle, disposable, blue, SteriPlast®
Leverandør: BURKLE
5378-5023
331-0360EA 700 NOK
331-0360
Ladle, long handle, disposable, blue, SteriPlast®
Øser
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work.
  • PS, blue
  • Individually packaged and sterilised, for single use
  • Reaches into every corner, handle length 300 mm
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Especially for the