Food palette knife spatula, SteriPlast®
Leverandør: BURKLE
5378-3035
331-0370EA 1825 NOK
331-0370
Food palette knife spatula, SteriPlast®
Spatler
The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle’s volumetric and dosing spoons.
  • PS, blue
  • Individually packaged and sterilised, for single use
  • L×W 192×20 mm
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Clean room manufactured (