Palette knife spatula, detectable, blue, SteriPlast®
Leverandør: BURKLE
5378-2035
331-0371EA 2250 NOK
331-0371
Palette knife spatula, detectable, blue, SteriPlast®
Spatler
The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle’s volumetric and dosing spoons.
  • PS detectable, blue
  • Individually packaged and sterilised, for single use
  • L×W 192×20 mm
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Can be detected by colourscanners, metal detectors or X-rays
  • Clean room manufactured (