Prøvetaker for væsker, LiquiSampler
Leverandør: BURKLE
5330-2100 5330-2200 5330-1100 5330-1200 5330-1060 5330-2060
331-2150EA 8000 NOK
331-2150 331-2151 331-2152 331-2153 331-0036 331-0037
Prøvetaker for væsker, LiquiSampler
Prøvetakere Væskeprøvetakere
Laget av ultra-ren og kjemisk inert PTFE/FEP eller PP for forurensningsfri prøvetaking. Passer til alle beholder og fat med åpninger med en Ø på minst 32 mm.
  • Taking apart and cleaning is extremely simple
  • One hand push button operation
  • Easy to clean − Surfaces are free of pores and crevices, preventing accumulation of dirt
  • Colour coded push buttons in red, blue, green, yellow, white and black
  • Round screw threads proven in foodstuffs hygiene
Suitable for point sampling, cross-sectional sampling, bottom sampling from open and closed barrels, vats, tanks,More Product Information