VWR®, Water Sampling Bottles
Leverandør: VWR Collection
331-3411EA 0 NOK
331-3411
VWR®, Water Sampling Bottles
Flasker Vannprøveflasker
HDPE.
  • Dosage in compliance with standards: ISO 5667-3/ISO 19458
  • Sodium thiosulphate in powder form
  • Tamper-evident red screw cap guarantees the sterility of the bottles prior to use
  • Shelf life up to 2 years
These bottles are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken. Square or rectangular bottles for more efficient storage.