Combustion trays, ceramic, AL23
Leverandør: Thermo Fisher Scientific
33183.KF 33185.KF 33186.KF
33183.KFEA 1275 NOK
33183.KF 33185.KF 33186.KF
Combustion trays, ceramic, AL23
Skip Forbrenningsskip
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.