Combustion trays, ceramic, AL24
Leverandør: Thermo Fisher Scientific
33187.KF
33187.KFEA 6475 NOK
33187.KF
Combustion trays, ceramic, AL24
Skip Forbrenningsskip
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.