Glassballer
Leverandør: VWR Chemicals
332124GEA 1550 NOK
332124G
Glassballer
Kuler Glasskuler
Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.