Supelco® PESTANAL® Organic Reference Standard, Triazoxid

Leverandør: Merck

33373-100MG
33373-100MGEA 670 NOK
33373-100MG
Supelco® PESTANAL® Organic Reference Standard, Triazoxid
Standarder Organiske standarder

  • GHS signal: GHS06


NB: P261 - P264 - P270 - P271 - P301 + P310 - P304 + P340 + P311

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6c1391959f43d94b5cee8b269c817205ce8cb94&vid=e6c1391959f43d94b5cee8b269c817205ce8cb94&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 72459-58-6
MDL No. MFCD00145438
Molekylvekt 247,64
Smeltepunkt 178...182 °C
Hardhetsklasse beskrivelse H301 + H331
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR