di-Natriumdisulfitt (≥58,5% SO₂)

Leverandør: Thermo Fisher Scientific
Danger

33375.36 33375.A3
33375.36EA 505 NOK
33375.36 33375.A3
di-Natriumdisulfitt (≥58,5% SO₂)
di-Natriumdisulfitt

CAS No.: 7681-57-4 Molecular Formula: Na2S2O5 Formula Weight: 190.10 Physical Form: Granular Density: 1.480 Sensitivity: Air and Moisture Sensitive Appearance: White to pale yellow Odour: Like rotten eggs pH: 3.5-5.0 Melting Point: 300° Solubility: Freely soluble in water, glycerol. Slightly soluble in alcohol Merck Reference: 14,8638 MDL No.: MFCD00167602

Formel: Na₂S₂O₅
Smeltepunkt: >300 °C
Tetthet: 1,480
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167602
CAS-nummer: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Indeks: 14,8638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR