Food standards, Fusarenon X, neat, Supelco®
Leverandør: Merck
33438-5MG
33438-5MGEA 9500 NOK
33438-5MG
Food standards, Fusarenon X, neat, Supelco®
Standarder Mat standarder
Fusarenon X is a trichothecene mycotoxin isolated from Fusarium nivale.
Fusarenon X may be used as a reference standard for the determination of fusarenon X in barley, wheat and corn samples by high-performance thin-layer chromatography (HPTLC).
NB: Storage temperature: Freezer −20 °C
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=764215d94f8c297d07032a245edb35da5fa2d016&vid=764215d94f8c297d07032a245edb35da5fa2d016&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Beskrivelse Fusarenon X
CAS-nr 23255-69-8
M-Clarity quality level MQ100