Ammoniumkarbonat ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

3352-20EA 0 NOK
3352-20
Ammoniumkarbonat ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™
Ammoniumkarbonat
Formel: (NH₄)₂CO₃
Smeltepunkt: 58 °C
Tetthet: 1,5 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010890
CAS-nummer: 10361-29-2
EINECS: 208-058-0
Merck Indeks: 13,00508

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 30 %
ACS - Assay (as NH₃) ≥30 %
ACS - Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Nonvolatile Matter ≤0.01 %
ACS - Sulfur Compounds (as SO₄) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR