Supelco® Organic Reference Standard, 4-Tert-Octylphenol-3,5-D₂ Monoethoxylate Solution

Leverandør: Merck

33523-1ML
33523-1MLEA 4125 NOK
33523-1ML
Supelco® Organic Reference Standard, 4-Tert-Octylphenol-3,5-D₂ Monoethoxylate Solution
Standarder Organiske standarder

  • GHS signal: GHS02, GHS07


NB: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dff45774aba69c2ec15d08f4162fac7410c026a&vid=2dff45774aba69c2ec15d08f4162fac7410c026a&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy-3,5-d₂]ethanol; 4-tert-OP1EO-D2
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 1173022-57-5
MDL No. MFCD09842825
Formel C₁₆D₂H₂₄O₂
Molekylvekt 252,39
Hardhetsklasse beskrivelse H225 - H319 - H336
Flammepunkt –17 °C - closed cup
Fraktforhold STD
Lagringsforhold –20 °C
Lagringstemp −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR