tri-Natriumcitrat dihydrat ≥99%, Gen-Apex for molekylærbiologi

Leverandør: VWR Chemicals

33615.268EA 975 NOK
33615.268
tri-Natriumcitrat dihydrat ≥99%, Gen-Apex for molekylærbiologi
tri-Natriumcitrat dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Smeltepunkt: 150 °C
Tetthet: 1,76 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150031
CAS-nummer: 6132-04-3
EINECS: 200-675-3
REACH: 01-2119457027-40
Merck Indeks: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Colouration (0,1 mol/l; water) Max. 10 APHA
Heavy metals (as Pb) Max. 2 ppm
Transmittance (260 nm) Min. 98 %
Transmittance (280 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR