Hydroxynaphthol blue indicator for metal titration, Fluka™

Leverandør: Honeywell Chemicals

Synonymer: 1-(2-Hydroxy-4-sulpho-1-naphthylazo)-2-naphthol-3,6-disulphonic acid trisodium salt

33936-10G 33936-50G
33936-10GEA 1550 NOK
33936-10G 33936-50G
Hydroxynaphthol blue indicator for metal titration, Fluka™
Hydroxynaphthol blue
Formel: C₂₀H₁₁N₂Na₃O₁₁S₃
MW: 620.47 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036393
CAS-nummer: 63451-35-4
EINECS: 264-197-7

Bestill nå

Spesifikasjon Testresultater

Identity (LC-MS) complying
Absorption maximum 640 - 650 nm
Absorbtion (1%/1cm) Min. 325
Suitability for Complexometry complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR