Pesticide reference standard, Febantel, VETRANAL(TM), analytical standard, Supelco®
Leverandør: Merck
33981-100MG-R
33981-100MG-REA 1375 NOK
33981-100MG-R
Pesticide reference standard, Febantel, VETRANAL(TM), analytical standard, Supelco®
Standarder Organiske standarder
Febantel might be utilised as a source of reference standard in the determination of febantel in samples of animal feed and pharmaceutical formulations, using high performance liquid chromatography (HPLC) and high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS-MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9963d00cd4343f814b8503b34015db991a0e1929&vid=f895e233a5178abac97bd9bc40a4d019bce6ab85&prid=web&clid=SAPDAM