Supelco® PESTANAL® Organic Reference Standard, Isoproturon-D₆

Leverandør: Merck

34017-10MG-R
34017-10MG-REA 2150 NOK
34017-10MG-R
Supelco® PESTANAL® Organic Reference Standard, Isoproturon-D₆
Standarder Organiske standarder

  • GHS signal: GHS08, GHS07, GHS09


NB: P202 - P260 - P273 - P280 - P308 + P313 - P391

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df7c86030bac8382eb70b5263a408ba33a3a2dcc&vid=136b8548858e377a5adaf4ec04898f2b47eab596&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 3-(4-Isopropylphenyl)-1,1-dimethylurea-d₆
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 217487-17-7
MDL No. MFCD01632296
Formel C₁₂H₁₂D₆N₂O
Molekylvekt 212,32
Hardhetsklasse beskrivelse H351 - H373 - H410
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR