Silikagel, BAKER ANALYZED® (4-10 netting) desiccant, with moisture indicator, J.T.Baker®

Leverandør: Avantor
Danger

3402-01EA 2425 NOK
3402-01
Silikagel, BAKER ANALYZED® (4-10 netting) desiccant, with moisture indicator, J.T.Baker®
Silikagel

Fargeendring:
Blå.................Aktivert
Fiolett..........10 % fuktighet absorbert
Rosa..........19 % fuktighet absorbert
Blekrosa..........28 % fuktighet absorbert

Formel: SiO₂
Tetthet: 2,10 kg/m³
MDL nummer: MFCD00011232
CAS-nummer: 63231-67-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR