Supelco® Organic Reference Standard, BDE No 138 Solution

Leverandør: Merck

34122-1ML
34122-1MLEA 4600 NOK
34122-1ML
Supelco® Organic Reference Standard, BDE No 138 Solution
Standarder Organiske standarder

  • GHS signal: GHS02, GHS08, GHS07, GHS09


NB: P210 - P273 - P301 + P310 + P331 - P302 + P352

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a18a5a9426c821f3b5b7b8bc1b0d681d1f06e25d&vid=05d300afbe0f7e961ce848c07593997e8f8d1e12&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 2,2′,3,4,4′,5′-Hexabromodiphenyl ether solution; 2,2′,3,4,4′,5′-HexaBDE; PBDE 138
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 182677-30-1
MDL No. MFCD08460508
Formel C₁₂H₄Br₆O
Molekylvekt 643,58
Hardhetsklasse beskrivelse H225 - H304 - H315 - H336 - H410
Flammepunkt –12 °C - closed cup
Fraktforhold STD
Lagringsforhold 2...8 °C
Lagringstemp 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR