Pesticide reference standard, Ronidazole-d3, VETRANAL(TM), analytical standard, Supelco®
Leverandør: Merck
34217-10MG
34217-10MGEA 5050 NOK
34217-10MG
Pesticide reference standard, Ronidazole-d3, VETRANAL(TM), analytical standard, Supelco®
Standarder Organiske standarder
Ronidazole-d₃ may be used internal standard in the determination of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue using ultra-high performance liquid chromatography coupled with mass spectrometry (UHPLC/MS-MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46f80e34b69569000fb6c19386f11cb781faa84e&vid=46f80e34b69569000fb6c19386f11cb781faa84e&prid=web&clid=SAPDAM