Perchloric acid 1.0 M HClO4 (1.0N) ≥98% volumetric solution, Fluka™

Leverandør: Honeywell Chemicals
Warning

34288-1L
34288-1LEA 1500 NOK
34288-1L
Perchloric acid 1.0 M HClO4 (1.0N) ≥98% volumetric solution, Fluka™
Perchloric acid
Formel: HClO₄
Tetthet: 1,054 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR