Bromfenolblått 0,04% in ethanol, TECHNICAL

Leverandør: VWR Chemicals
Danger

Synonymer: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

34307.237EA 564 NOK
34307.237
Bromfenolblått 0,04% in ethanol, TECHNICAL
Bromfenolblått

  • Transition range: pH 3.0 - 4.6

Formel: C₁₉H₁₀Br₄O₅S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005875
CAS-nummer: 115-39-9
EINECS: 204-086-2
FN: 1170
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR