Silikagel Grade 22 pore size 60 Å, 60-200 mesh, Supelco®

Leverandør: Merck

343323-1KG
343323-1KGEA 3750 NOK
343323-1KG
Silikagel Grade 22 pore size 60 Å, 60-200 mesh, Supelco®
Silikagel
Formel: SiO₂
Kokepunkt: 2230 °C
Smeltepunkt: > 1600 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da46472b9dc6418bdb67b3a7a4103fe065e14a71&vid=5e87d07d41a322ec74f287e3802cc81315110215&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR