n-Heksan ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Leverandør: Honeywell Chemicals
Danger
Synonymer: Hexyl hydride, Hexanen, Hexane
34484-7L 34484-18L 34484-45L 34484-1L 34484-2.5L 34484-4X2.5L
34484-7LEA 7375 NOK
34484-7L 34484-18L 34484-45L 34484-1L 34484-2.5L 34484-4X2.5L
n-Heksan ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
n-Heksan
  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Formel: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Kokepunkt: 66…69 °C(1.013 hPa)
Smeltepunkt: –95 °C
Tetthet: 0.660…0.668 g/cm³ (20 °C)
Flash punkt: 7.4 %(V)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009520
CAS-nummer: 110-54-3
EINECS: 203-777-6
FN: 1208
ADR: 3 II
Merck Indeks: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Assay (GC) Min. 95 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Autoignition Temperature 453 °F
Explosion Limit 7.7 %
Vapor Pressure ~132 mmHg ( 20 °C)
Determination of P/N ≤5 ng/L