Idranal® III 99-101% (by HPLC) Reag. Ph. Eur. for complexometry (NTA ≤0.1%), Fluka™

Leverandør: Honeywell Chemicals
Warning

34549-25KG 34549-100G 34549-1KG 34549-6X1KG 34549-500G
34549-25KGEA 35600 NOK
34549-25KG 34549-100G 34549-1KG 34549-6X1KG 34549-500G
Idranal® III 99-101% (by HPLC) Reag. Ph. Eur. for complexometry (NTA ≤0.1%), Fluka™
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 244…250 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 101.0 %
Loss on drying (150°C, 3h) Max. 10 %
pH (5 %, 20°C) 4.0 - 5
pH (5 %, 25 °C) 4.0 - 6.0
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0002 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.005 %
Nitrilotriacetic acid (HPLC) Max. 0.1 %
Water insoluble matter Max. 0.005 %
Appearance of the solution complying
Identity (IR) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR