Metanol, dry, Hydranal™ medium for volumetric one-component Karl Fischer titration, Fluka™

Leverandør: Honeywell Chemicals
Danger

Synonymer: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

34741-1L 34741-2.5L 34741-6X1L 34741-4X2.5L
34741-1LEA 320 NOK
34741-1L 34741-2.5L 34741-6X1L 34741-4X2.5L
Metanol, dry, Hydranal™ medium for volumetric one-component Karl Fischer titration, Fluka™
Metanol

Suitability: use tested, Reagent for volumetric one-component KF titration (working medium)

Formel: CH₃OH
MW: 32.04 g/mol
Kokepunkt: 64…65 °C (1.013 hPa)
Smeltepunkt: –98 °C
Tetthet: 0.79 g/cm³ (20 °C)
Flash punkt: 11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004595
CAS-nummer: 67-56-1
EINECS: 200-659-6
FN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Indeks: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Suitability (for KF-titration) test passed
Water (Karl Fischer) Max. 0.01 %
Number Of Samples Min. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR