Coulomat oils for Karl Fischer titration, HYDRANAL™
Leverandør: Honeywell Chemicals
34868-500ML 34868-100ML 34868-6X100ML 34868-6X500ML
34868-500MLEA 1750 NOK
34868-500ML 34868-100ML 34868-6X100ML 34868-6X500ML
Coulomat oils for Karl Fischer titration, HYDRANAL™
Karl Fischer titranter og reagenser
Coulomat oil is a reagent for coulometric KF titration in oils (anolyte solution), preferred to use for the cells with diaphragm.