Acetonitril med 0,1% (v/v) Trifluoreddiksyre ≥99.5%, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Danger

34976-2.5L
34976-2.5LEA 3450 NOK
34976-2.5L
Acetonitril med 0,1% (v/v) Trifluoreddiksyre ≥99.5%, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Acetonitril med 0,1% (v/v) Trifluoreddiksyre

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities


RTECS Number: 200-835-2

Kokepunkt: 81…82 °C
Tetthet: 0,78 g/cm³ (20 °C)
Flash punkt: 6 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001878
CAS-nummer: 76-05-1
EINECS: 200-835-2
FN: 1648
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Assay (GC) Min. 99.5 %
Assay trifluoroacetic acid 0.093 - 0.107 Vol.%
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) max. 2 ppm
Transmittance at 210 nm min. 30 %
Transmittance at 230 nm min. 50 %
Transmittance from 254 nm min. 90 %
HPLC Gradient at 254 nm max. 50 mAU
LC-MS application complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR