Natriumhydrogenkarbonat 99.0-100.5% (dried basis), pulver USP, FCC, J.T.Baker®

Leverandør: Avantor

3509-01EA 665 NOK
3509-01 3509-05 3509-07 3509-09
Natriumhydrogenkarbonat 99.0-100.5% (dried basis), pulver USP, FCC, J.T.Baker®
Natriumhydrogenkarbonat

No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Sertifiseringer: Meets USP and FCC requirements.


NB: Not for use in hemodialysis.

Formel: NaHCO₃
Smeltepunkt: 270 °C
Tetthet: 2,20 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003528
CAS-nummer: 144-55-8
EINECS: 205-633-8
Merck Indeks: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets F.C.C. Requirements Passes Test
Meets U.S.P Requirements ≤0.015%
Food GMP Manufactured Product ≤0.25%
GMP Manufactured Product Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤20 ppm
Bulk Food Chemical ≤0.015%
USP - Identification Passes Test
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Insoluble Substances Passes Test
USP - Normal Carbonate Passes Test
USP - Limit of Ammonia ≤20 ppm
USP - Chloride (Cl) ≤0.015 %
USP - Arsenic (As) ≤2 ppm
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
FCC - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
Appearance (White powder) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
This product is not intended for use in hemodialysis.
The net weight of the product in each container is within +/- 2% of the
netweight stated on the label.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR