Ammonium sulphide 20 - 24% vandig løsning, AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

3524-04EA 2875 NOK
3524-04
Ammonium sulphide 20 - 24% vandig løsning, AR®, Macron Fine Chemicals™
Ammonium sulphide
Formel: H₈N₂S
MW: 68,14 g/mol
Tetthet: 1,00 g/cm³
MDL nummer: MFCD00010892
CAS-nummer: 12135-76-1
EINECS: 235-223-4
FN: 2683
ADR: 8 (6.1) (3),II

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use 20.0 - 24.0%
Assay (as (NH₄)₂S) 20.0 - 24.0 %
Antimony (Sb) ≤0.0005 %
Arsenic (As) ≤0.0001 %
Carbonate (CO₃) ≤0.005 %
Chloride (Cl) ≤0.005 %
Residue after Ignition ≤0.04 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR