Multi Ion anion IC standard solution, Specpure® (Waters) in H₂O

Leverandør: Thermo Fisher Scientific

35564.AEEA 4125 NOK
35564.AE
Multi Ion anion IC standard solution, Specpure® (Waters) in H₂O
Standarder Anionstandarder Multi-Element Anioinic Standards

Physical Form: Liquid
MDL No.: MFCD00144814

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR