Multi Ion anion IC standard solution, Specpure® (Dionex) in H₂O

Leverandør: Thermo Fisher Scientific

35565.AEEA 3775 NOK
35565.AE
Multi Ion anion IC standard solution, Specpure® (Dionex) in H₂O
Standarder Anionstandarder Multi-Element Anioinic Standards

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00144813

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00144813
Fyskisk form Liquid
Lukt Odourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR