Kaliumdikromat standardised solution 0.025 N vandig løsning

Leverandør: Thermo Fisher Scientific
Danger

35589.AP 35589.K2
35589.APEA 410 NOK
35589.AP 35589.K2
Kaliumdikromat standardised solution 0.025 N vandig løsning
Kaliumdikromat

CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid MDL No.: MFCD00011367

Formel: K₂Cr₂O₇
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011367
CAS-nummer: 7778-50-9
EINECS: 231-906-6
FN: 3287
ADR: 6.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR