Oksalsyre (Etandisyre) 10% (w/v) vandig løsning

Leverandør: Thermo Fisher Scientific
Danger

35617.K2 35617.K7
35617.K2EA 890 NOK
35617.K2 35617.K7
Oksalsyre (Etandisyre) 10% (w/v) vandig løsning
Oksalsyre (Etandisyre)

Molecular Formula: HO2CCO2H Formula Weight: 90.04 Physical Form: Liquid Odour: Odourless UN No.: 3265 MDL No.: MFCD00002573

Formel: C₂H₂O₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002573
CAS-nummer: 144-62-7
EINECS: 205-634-3
FN: 3265
ADR: 8,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR