Glycerin, Calbiochem®

Leverandør: Merck Millipore Calbiochem

Synonymer: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

356352-1
356352-1EA 3625 NOK
356352-1
Glycerin, Calbiochem®
Glycerin
Formel: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Kokepunkt: 290
Smeltepunkt: 18.6
Tetthet: 1.26 g/cm³ (20 °C)
Flash punkt: 177 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004722
CAS-nummer: 56-81-5
EINECS: 200-289-5
Merck Indeks: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR